Learn More
Succinophenone (Diphenacyl), Mikromol™
Product Code. 30050489
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050489 25 mg 25mg
30049383 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30050489 Supplier Mikromol Supplier No. MM0039.010025
 Restricted Item
Forventet forsendelse: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Succinophenone (Diphenacyl)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Succinophenone (Diphenacyl)
CAS 495-71-6
Analyte or Component Names Succinophenone (Diphenacyl)
Shipping Condition Room Temperature
Molecular Formula C16 H14 O2
Certifications/Compliance ISO 9001
Synonym Succinophenone, Diphenacyl
InChI Formula InChI=1S/C16H14O2/c17-15(13-7-3-1-4-8-13)11-12-16(18)14-9-5-2-6-10-14/h1-10H,11-12H2
SMILES O=C(CCC(=O)c1ccccc1)c2ccccc2
IUPAC Name 1,4-diphenylbutane-1,4-dione
Molecular Weight (g/mol) 238.28
Quantity 25 mg
Formula Weight 238.0994 g/mol