Få mere at vide
Succinimidyl Nervonate, TRC
Artikelnummer. 30412350
Skift visning
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30412350 5 mg
30412753 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30412350 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6909505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Succinimidyl Nervonate
CAS 191354-88-8
Molekylær formel C28 H49 N O4
Mængde 5 mg
InChI formel InChI=1S/C28H49NO4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-28(32)33-29-26(30)24-25-27(29)31/h9-10H,2-8,11-25H2,1H3/b10-9-
SMIL CCCCCCCC\C=C/CCCCCCCCCCCCCC(=O)ON1C(=O)CCC1=O
IUPAC navn (2,5-dioxopyrrolidin-1-yl) (Z)-tetracos-15-enoate
Molekylvægt (g/mol) 463.69
Formel vægt 463.3662