Få mere at vide
Succinic Anhydride-D4, TRC
Artikelnummer. 30417773
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30417773 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30417773 Leverandør Toronto Research Chemicals Leverandørnr. TRCS688773500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Succinic Anhydride-d4
CAS 14341-86-7
Molekylær formel C4 2H4 O3
Mængde 500 mg
Synonym 2,5-Furandione-3,4-d2','dihydro-d2- (9CI','ACI), Dihydro-d2-2,5-furandione-3,4-d2 (ACI), Succinic-d4 anhydride (7CI','8CI), 3,3,4,4-Tetradeuteriooxolane-2,5-dione, Succinic-2,2,3,3-d4 anhydride, Succinic Anhydride-D4
InChI formel InChI=1S/C4H4O3/c5-3-1-2-4(6)7-3/h1-2H2/i1D2,2D2
SMIL [2H]C1([2H])C(=O)OC(=O)C1([2H])[2H]
IUPAC navn 3,3,4,4-tetradeuteriooxolane-2,5-dione
Molekylvægt (g/mol) 104.1
Formel vægt 104.0412