Få mere at vide
DiscoverX Succinic acid disodium salt
Artikelnummer. 17322519
Encompass
Skift visning
10 mg
Artikelnummer.
17322519 10 mg
1 options
Artikelnummer. 17322519 Leverandør DiscoverX Leverandørnr. 921193
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Succinic acid disodium salt is an agonist for GPR91 (SUCNR1). Provided as 10mg of dry powder.
Succinic acid is a dicarboxylic acid which takes the form of succinate in living organisms. Succinate is produced in inflammatory conditions, may be associated with rheumatoid arthritis, and its accumulation is associated with tumorigenesis. It acts through the succinate receptor, SUCNR1 (or GPR91), leading to enhanced IL-1β production. Provided as 10mg of dry powder.