Få mere at vide
Succinic Acid-d6, CDN
Artikelnummer. 30321465
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30321465 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321465 Leverandør CDN Leverandørnr. CDND12001G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Succinic-d6 Acid
CAS 21668-90-6
Molekylær formel C4 D6 O4
Mængde 1 g
Synonym Succinic Acid-d6, Butanedioic-2,2,3,3-d4 acid-1,3-d2 (ACI), Butanedioic-d4 acid-d2 (9 CI), Succinic-d4 acid-d2 (6 CI, 7 CI, 8 CI)
InChI formel InChI=1 S/C4H6O4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)/i1D2,2D2/hD2
SMIL [2 H]OC(=O)C([2 H])([2 H])C([2 H])([2 H])C(=O)O[2 H]
IUPAC navn dideuterio 2,2,3,3-tetradeuteriobutanedioate
Molekylvægt (g/mol) 124.13
Formel vægt 124.0643 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity