Få mere at vide
Succinic Acid-13C4, TRC
Artikelnummer. 30134350
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134350 25 mg
30003390 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134350 Leverandør Toronto Research Chemicals Leverandørnr. TRCS68876725MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 201595-67-7
Molekylær formel 13C4H6O4
Molekylvægt (g/mol) 122.06
Synonym Butanedioic Acid-13C4, 1,2-Ethanedicarboxylic Acid-13C4, 1,4-Butanedioic Acid-13C4, A 12084-13C4, Amber Acid-13C4, Asuccin-13C4, Dihydrofumaric Acid-13C4, Katasuccin-13C4, NSC 106449-13C4, NSC 25949-13C4, Wormwood Acid-13C4, Yantar-antitox-13C4;
SMIL O=[13C](O)[13C]([HH])[13C]([HH])[13C](O)=O
Tekniske data
Kemisk navn eller materiale Succinic Acid-13C4
Mængde 25 mg
InChI formel InChI=1S/C4H6O4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)/i1+1,2+1,3+1,4+1