Få mere at vide
MedChemExpress Suc-Leu-Leu-Val-Tyr-AMC
Artikelnummer. 16492789
Encompass_Preferred
Skift visning
Mængde:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16492789 1 mg
16401998 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16492789 Leverandør MedChemExpress Leverandørnr. HYP1002
MedChemExpress Suc-Leu-Leu-Val-Tyr-AMC is a fluorogenic substrate.
Tekniske data
Mængde 1 mg
Indhold og opbevaring Protect from light, stored under nitrogen ∣ Powder -80°C 2 years, -20°C 1 year ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light, stored under nitrogen)
CAS 94367-21-2
Opløselighed DMSO : ≥ 20 mg/mL (26.18 mM)
Produkttype Dye Reagents
Molekylvægt (g/mol) 763.88 g/mol
Forsendelsestilstand Room temperature
Mål NO Synthase ∣ MMP ∣ Endogenous Metabolite