Få mere at vide
MedChemExpress Substance P (7-11)
Artikelnummer. 16494056
Encompass_Preferred
Skift visning
Mængde:
25 mg
Pakningsstørrelse:
25mg
Artikelnummer. Mængde unitSize
16494056 25 mg 25mg
1 options
Artikelnummer. 16494056 Leverandør MedChemExpress Leverandørnr. HYP1492
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
MedChemExpress Substance P (7-11) is a C-terminal fragment of Substance P which can cause an increase in the intracellular calcium concentration.
Tekniske data
CAS 51165-05-0
Indhold og opbevaring Please store the product under the recommended conditions in the Certificate of Analysis.
Til brug med (applikation) Metabolism-protein/nucleotide metabolism
Molekylvægt (g/mol) 612.78 g/mol
Produkttype Peptides