Få mere at vide
Suberoyl Bis-hydroxamic Acid, TRC
Artikelnummer. 30410429
Skift visning
Mængde:
100 mg
1 g
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30410429 100 mg -
30417717 1 g -
30420897 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30410429 Leverandør Toronto Research Chemicals Leverandørnr. TRCS688750100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 38937-66-5
Molekylær formel C8 H16 N2 O4
Molekylvægt (g/mol) 204.22
IUPAC navn N,N'-dihydroxyoctanediamide
SMIL ONC(=O)CCCCCCC(=O)NO
Tekniske data
Kemisk navn eller materiale Suberoyl Bis-hydroxamic Acid
Mængde 100 mg
InChI formel InChI=1S/C8H16N2O4/c11-7(9-13)5-3-1-2-4-6-8(12)10-14/h13-14H,1-6H2,(H,9,11)(H,10,12)
Formel vægt 204.111