Få mere at vide
Suberanilic Acid, TRC
Artikelnummer. 30417258
Skift visning
Mængde:
100 mg
200 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30417258 200 mg
30414345 100 mg
30426101 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30417258 Leverandør Toronto Research Chemicals Leverandørnr. TRCS688650200MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 149648-52-2
Molekylær formel C14 H19 N O3
Molekylvægt (g/mol) 249.31
Synonym Octanoic acid, 8-oxo-8-(phenylamino)-, 8-Oxo-8-(phenylamino)octanoic acid, Suberanilic acid
IUPAC navn 8-anilino-8-oxooctanoic acid
SMIL OC(=O)CCCCCCC(=O)Nc1ccccc1
Tekniske data
Kemisk navn eller materiale Suberanilic Acid
Mængde 200 mg
InChI formel InChI=1S/C14H19NO3/c16-13(15-12-8-4-3-5-9-12)10-6-1-2-7-11-14(17)18/h3-5,8-9H,1-2,6-7,10-11H2,(H,15,16)(H,17,18)
Formel vægt 249.1365