missing translation for 'onlineSavingsMsg'
Få mere at vide

STEMCELL Technologies SU5416​

Artikelnummer. 17170222
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17170222 50 mg
17160222 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17170222 Leverandør STEMCELL Technologies Leverandørnr. 73444

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Tyrosine kinase inhibitor; Inhibits KDR, PDGFR, KIT, RET, FLT-3, ABL, and ALK

  • SU5416 is a tyrosine kinase inhibitor best known as an ATP-competitive inhibitor of the kinase insert domain (KDR/VEGFR2/FLK1)
  • In addition to inhibiting KDR (IC50 = 1 μM), SU5416 also inhibits PDGFR (IC50 = 20 μM), KIT (IC50 = 30 nM), RET (IC50 = 170 nM), FLT-3 (IC50 = 160 nM), ABL (IC50 = 11 μM), and ALK (IC50 = 1.2 μM)
  • SU5416 does not inhibit EGFR or FGFR tyrosine kinases (IC50 > 100 μM; Fong et al.; Litz; Mologni et al.)
  • Prevents angiogenesis, thereby inhibiting tumor growth, catalysis and vascularization for a variety of cancers (Litz; Fong et al.)
  • Inhibits RET-mediated transformation of NIH-3T3 mouse fibroblasts and Ba/F3 mouse pro-B cells (Mologni et al.)
  • Causes pulmonary hypertension in SuHx rat model of pulmonary arterial hypertension, when combined with hypoxia (de Raaf et al.; Mizuno et al.)
  • Inhibits TGFβ1 activation and delays wound healing in rats (Haroon et al.)

Tekniske data

CAS 204005-46-9
Synonym NSC 696819; Semaxinib; Sugen 5416; VEGFR 2 Kinase Inhibitor
Mængde 50 mg
Molekylær formel C15H14N2O
Renhed ≥98%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.