missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
STEMCELL Technologies SU5416
Beskrivelse
- SU5416 is a tyrosine kinase inhibitor best known as an ATP-competitive inhibitor of the kinase insert domain (KDR/VEGFR2/FLK1)
- In addition to inhibiting KDR (IC50 = 1 μM), SU5416 also inhibits PDGFR (IC50 = 20 μM), KIT (IC50 = 30 nM), RET (IC50 = 170 nM), FLT-3 (IC50 = 160 nM), ABL (IC50 = 11 μM), and ALK (IC50 = 1.2 μM)
- SU5416 does not inhibit EGFR or FGFR tyrosine kinases (IC50 > 100 μM; Fong et al.; Litz; Mologni et al.)
- Prevents angiogenesis, thereby inhibiting tumor growth, catalysis and vascularization for a variety of cancers (Litz; Fong et al.)
- Inhibits RET-mediated transformation of NIH-3T3 mouse fibroblasts and Ba/F3 mouse pro-B cells (Mologni et al.)
- Causes pulmonary hypertension in SuHx rat model of pulmonary arterial hypertension, when combined with hypoxia (de Raaf et al.; Mizuno et al.)
- Inhibits TGFβ1 activation and delays wound healing in rats (Haroon et al.)
Tekniske data
Tekniske data
| CAS | 204005-46-9 |
| Synonym | NSC 696819; Semaxinib; Sugen 5416; VEGFR 2 Kinase Inhibitor |
| Mængde | 10 mg |
| Molekylær formel | C15H14N2O |
| Renhed | ≥98% |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?