missing translation for 'onlineSavingsMsg'
Få mere at vide

STEMCELL Technologies SU11274

Artikelnummer. 17140222
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17140222 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17140222 Leverandør STEMCELL Technologies Leverandørnr. 73432

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

HGF pathway inhibitor; Inhibits MET

  • SU11274 is a selective, ATP-competitive inhibitor of MET receptor tyrosine kinase (IC50 = 20 nM; Sattler et al.) It shows good selectivity towards MET versus other receptor tyrosine kinases with IC50 values of 1.3 μM and 9.7 μM for kinase insert domain receptor (KDR) and fibroblast growth factor receptor-1 (FGFR1), respectively (Sattler et al.) It retains activity for certain MET mutants, for example H1112Y and M1268T (Berthou et al.)
  • Induces apoptosis and cell cycle arrest in TPR-MET-transformed mouse pre-B (Ba/F3) cells (Sattler et al.)
  • Inhibits cell viability and motility of human head and neck squamous cell carcinoma cell lines in vitro (Seiwert et al.)
  • Inhibits cell viability of c-MET-expressing human non-small cell lung carcinoma cell lines in vitro (Ma et al.)

Tekniske data

CAS 658084-23-2
Synonym Met Kinase Inhibitor
Mængde 1 mg
Molekylær formel C28H30ClN5O4S
Renhed ≥98%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.