Få mere at vide
SU 5402, MedChemExpress
Artikelnummer. 16493948
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16493948 10 mg -
16484966 100 mg -
16476667 5 mg 5mg
16450307 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16493948 Leverandør Medchem Express Leverandørnr. HY10407
MedChemExpress SU 5402 is a potent multi-targeted receptor tyrosine kinase inhibitor with IC50 of 20 nM, 30 nM, and 510 nM for VEGFR2, FGFR1, and PDGFRβ, respectively.
SU 5402 is a potent multi-targeted receptor tyrosine kinase inhibitor with IC50 of 20 nM, 30 nM, and 510 nM for VEGFR2, FGFR1, and PDGFRβ, respectively.
Tekniske data
Kemisk navn eller materiale SU 5402
Fysisk form Solid
CAS 215543-92-3
Farve Brown
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C17H16N2O3
Til brug med (applikation) Cancer-Kinase/protease
Opløselighedsinformation DMSO : ≥ 30 mg/mL (101.24 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMIL O=C(CCC1=C(NC=C1C)/C=C2C(NC3=C\2C=CC=C3)=O)O
Molekylvægt (g/mol) 296.32
Formel vægt 296.32
Procent renhed 96.71%