missing translation for 'onlineSavingsMsg'
Learn More

SU 1498, TRC

Product Code. 30413866
Change view
Click to view available options
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30413866 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413866 Supplier Toronto Research Chemicals Supplier No. TRCS687990250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (E)-2-cyano-3-[4-hydroxy-3,5-di(propan-2-yl)phenyl]-N-(3-phenylpropyl)prop-2-enamide
CAS 168835-82-3
Molekylær formel C25 H30 N2 O2
Mængde 250 mg
Synonym SU 1498, (E)-2-Cyano-3-[4-hydroxy-3,5-bis(1-methylethyl)phenyl]-N-(3-phenylpropyl)-2-propenamide, AG 1498, SU 1498, Tyrphostin SU 1498, (E)-2-Cyano-3-(4-hydroxy-3,5-diisopropylphenyl)-N-(3-phenylpropyl)acrylamide
InChI formel InChI=1S/C25H30N2O2/c1-17(2)22-14-20(15-23(18(3)4)24(22)28)13-21(16-26)25(29)27-12-8-11-19-9-6-5-7-10-19/h5-7,9-10,13-15,17-18,28H,8,11-12H2,1-4H3,(H,27,29)/b21-13+
SMIL CC(C)c1cc(\C=C(/C#N)\C(=O)NCCCc2ccccc2)cc(C(C)C)c1O
IUPAC navn (E)-2-cyano-3-[4-hydroxy-3,5-di(propan-2-yl)phenyl]-N-(3-phenylpropyl)prop-2-enamide
Molekylvægt (g/mol) 390.518
Formel vægt 390.231
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.