Få mere at vide
Styrene-alpha,beta,beta-d3, CDN
Artikelnummer. 30321009
Skift visning
Mængde:
0.5 g
Pakningsstørrelse:
0.5g
Artikelnummer. Mængde unitSize
30321009 0.5 g 0.5g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321009 Leverandør CDN Leverandørnr. CDND04580.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Styrene vinyl-d3
CAS 3814-93-5
Molekylær formel C8 2H3 H5
Mængde 0.5 g
Synonym Benzene, ethenyl-1,2,2-d3-, Benzene, ethenyl-d3- (9 CI), Styrene-α,β,β-d3 (6 CI,7 CI,8 CI), Ethenyl-1,2,2-d3benzene, α,β,β-Trideuterostyrene, Styrene (vinyl-D3), Styrene D3 (vinyl-D3)
InChI formel InChI=1 S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7 H,1H2/i1D2,2 D
SMIL [2 H]C(=C([2 H])c1ccccc1)[2 H]
IUPAC navn 1,2,2-trideuterioethenylbenzene
Molekylvægt (g/mol) 107.17
Formel vægt 107.0814 g/mol
Procent renhed 97 atom % D, min 98% Chemical Purity