Få mere at vide
Styrene-a-d1, TRC
Artikelnummer. 30421286
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30421286 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421286 Leverandør Toronto Research Chemicals Leverandørnr. TRCS68779150MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Styrene-alpha-d1
CAS 1193-80-2
Molekylær formel C8 2H H7
Mængde 50 mg
Synonym Benzene','ethenyl-1-d-, Styrene-α-d (6CI,7CI,8CI), Ethenyl-1-dbenzene, Styrene-α-d1, α-Deuteriostyrene, α-Deuterostyrene, Styrene-α-D1
InChI formel InChI=1S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7H,1H2/i2D
SMIL [2H]C(=C)c1ccccc1
IUPAC navn 1-deuterioethenylbenzene
Molekylvægt (g/mol) 105.16
Formel vægt 105.0689