Få mere at vide
Styrene-2,3,4,5,6-d5, CDN
Artikelnummer. 30321560
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321560 1 g -
30321146 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321560 Leverandør CDN Leverandørnr. CDND01261G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Styrene D5 (phenyl-D5)
CAS 5161-29-5
Molekylær formel C8 2H5 H3
Mængde 1 g
Synonym Benzene-1,2,3,4,5-d5, 6-ethenyl-, Benzene-d5, ethenyl- (9 CI), Styrene-2,3,4,5,6-d5 (7 CI,8 CI), 6-Ethenylbenzene-1,2,3,4,5-d5, 2,3,4,5,6-Pentadeuteriostyrene, Styrene (ring-D5), Styrene D5, Styrene D5 (phenyl-D5)
InChI formel InChI=1 S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7 H,1H2/i3D,4 D,5 D,6 D,7 D
SMIL [2 H]c1c([2 H])c([2 H])c(C=C)c([2 H])c1[2 H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-ethenylbenzene
Molekylvægt (g/mol) 109.18
Formel vægt 109.094 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity