Få mere at vide
Styramate, LoGiCal
Artikelnummer. 30460740
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30460740 10 mg
1 options
Artikelnummer. 30460740 Leverandør LoGiCal Leverandørnr. LGCFOR1676.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Styramate
CAS 94-35-9
Analyt- eller komponentnavne Styramate
Forsendelsestilstand Room Temperature
Molekylær formel C9 H11 N O3
Certificeringer/compliance ISO 9001
Synonym 1,2-Ethanediol, 1-phenyl-, 2-carbamate, 2-Hydroxy-2-phenylethyl carbamate, 2-Phenyl-2-hydroxyethyl carbamate, Linaxar, Myospaz, NSC 115889, Sinaxar, Styramat, Styramate, Carbamic acid, beta-hydroxyphenethyl ester (6CI,7CI), beta-Hydroxyphenethyl carbamate
InChI formel InChI=1S/C9H11NO3/c10-9(12)13-6-8(11)7-4-2-1-3-5-7/h1-5,8,11H,6H2,(H2,10,12)
Løsningstype Neat
SMIL NC(=O)OCC(O)c1ccccc1
IUPAC navn (2-hydroxy-2-phenylethyl) carbamate
Industry Type Clinical Testing
Molekylvægt (g/mol) 181.19
Mængde 10 mg
Formel vægt 181.0739 g/mol