Få mere at vide
(-)-Strigolactone GR24, TRC
Artikelnummer. 30011160
Skift visning
Mængde:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011160 5 mg
30003219 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011160 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6876105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 188062-52-4
Molekylær formel C17H14O5
Molekylvægt (g/mol) 298.29
Synonym (3E,3aS,8bR)-3-[[[(2S)-2,5-Dihydro-4-methyl-5-oxo-2-furanyl]oxy]methylene]-3,3a,4,8b-tetrahydro-2H-indeno[1,2-b]furan-2-one, [3aS-[3E(R*),3aalpha,8balpha]]-3-[[(2,5-Dihydro-4-methyl-5-oxo-2-furanyl)oxy]methylene]-3,3a,4,8b-tetrahydro-2H-indeno[1,2-b]furan-2-one;
IUPAC navn (3E,3aS,8bR)-3-[[(2S)-4-methyl-5-oxo-2H-furan-2-yl]oxymethylidene]-4,8b-dihydro-3aH-indeno[1,2-b]furan-2-one
SMIL CC1=C[C@@H](O\C=C\2/[C@@H]3Cc4ccccc4[C@@H]3OC2=O)OC1=O
Tekniske data
Kemisk navn eller materiale (-)-Strigolactone GR24
Mængde 5 mg
InChI formel InChI=1S/C17H14O5/c1-9-6-14(21-16(9)18)20-8-13-12-7-10-4-2-3-5-11(10)15(12)22-17(13)19/h2-6,8,12,14-15H,7H2,1H3/b13-8+/t12-,14-,15-/m0/s1
Formel vægt 298.0841