Få mere at vide
Kartell™ Stopper, conical, red rubber, upper\lower diameters 42 x 32mm, height 41mm
Kartell™ Red Rubber Conical Stoppers are suitable for high temperatures and are used in combination with a flask, bottle, or tube.
Brand:  Kartell™ 0114300
Produktkode 12617775
  • ONLINE EXCLUSIVE PRICE
    168.00 DKK
    159.43 DKK / Pakke med 5
    Save 8.57 DKK
    5% Slukket
Kun null tilbage
Forventet forsendelse: 11-02-2026
Tilføj til indkøbskurv
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Kartell™ Red Rubber Conical Stoppers are suitable for high temperatures and are used in combination with a flask, bottle, or tube.
  • For general purpose use
  • Red rubber
  • Conical shape
  • Size: lower (bottom) diameter x height