Learn More
Kartell™ Red Rubber Conical Stoppers
Kartell™ Red Rubber Conical Stoppers are suitable for high temperatures and are used in combination with a flask, bottle, or tube.
Brand:  Kartell™ 0114000
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10439782
  • 346.00 DKK / Pack of 25
Estimated Shipment
17-03-2021
Add to basket
Description
Description
  • For general purpose use
  • Red rubber
  • Conical shape
  • Size: lower (bottom) diameter x height
Specifications
Specifications
General purpose laboratory
25/Pk.
Red
Rubber
34mm
25 x 34mm
32mm
Conical