Learn More
Kartell™ Stopper, conical, red rubber, upper\lower diameters 26 x 19mm, height 28mm
Kartell™ Red Rubber Conical Stoppers are suitable for high temperatures and are used in combination with a flask, bottle, or tube.
Brand:  Kartell™ 0113800
Additional Details : Weight : 0.01000kg
Product Code. 10429782
  • 257.00 DKK / Pack of 25
Estimated Shipment: 10-05-2024
to see stock.
Description
Description
Kartell™ Red Rubber Conical Stoppers are suitable for high temperatures and are used in combination with a flask, bottle, or tube.
  • For general purpose use
  • Red rubber
  • Conical shape
  • Size: lower (bottom) diameter x height
Specifications
Specifications
Red
28mm
19mm
26mm
Rubber
General purpose laboratory
19 x 28mm
Conical Stopper
25/Pk.
Conical