missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ STOML1 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 13289088
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
Pakningsstørrelse:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
13289088 100 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 13289088 Leverandør Invitrogen™ Leverandørnr. PA521557

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Recommended positive controls: Molt-4. Predicted reactivity: Mouse (84%), Rat (83%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Stomatin-like protein 1 (STOML1) is encoded by the STOML1 gene located on chromosome 17 in humans. It is part of a family of five mammalian genes that encode peripheral membrane proteins involved in modulating ion channel functions. STOML1 is known to interact specifically with members of the acid-sensing ion channel (ASIC) family, influencing their proton sensitivity. These interactions help regulate ion transport and signaling pathways, which are critical for cellular homeostasis. In addition, STOML1 has been studied in the context of oral squamous cell carcinoma (OSCC), where it is often overexpressed along with STOML2, potentially affecting cancer progression. Structurally, STOML1 contains a stomatin domain which is crucial for its role in membrane association and ion channel modulation. Its expression in olfactory sensory neurons (OSNs) is comparatively lower than other family members, but it still contributes to the localization and functional integrity of related proteins in these cells.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen STOML1
Applikationer Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Koncentration 1 mg/mL
Konjugeret Unconjugated
Formulering 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene STOML1
Genadgangsnr. Q9UBI4
Gene Alias 1810015E19Rik; AI847431; EPB72-like 1; EPB72-like protein 1; hUNC-24; MSTP019; Protein unc-24 homolog; RGD1559463; SLP1; SLP-1; stomatin (EBP72)-like 1; stomatin (EPB72)-like 1; stomatin like 1; stomatin-like 1; stomatin-like protein 1; Stomatin-related protein; STOML1; STORP; UNC24; WPB72
Gensymboler STOML1
Værtsarter Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human STOML1. The exact sequence is proprietary.
Oprensningsmetode Antigen affinity chromatography
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 9399
Målarter Human
Indhold og opbevaring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.