missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
STK36 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-15339-100UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
STK36 Polyclonal antibody specifically detects STK36 in Human, Mouse samples. It is validated for Western Blot
Tekniske data
| STK36 | |
| Polyclonal | |
| PBS, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| EC 2.7.11, EC 2.7.11.1, Fused homolog, FUserine/threonine kinase 36, fused homolog, KIAA1278DKFZp434N0223, serine/threonine kinase 36, serine/threonine kinase 36 (fused homolog, Drosophila), serine/threonine-protein kinase 36 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 250-450 of human STK36 (NP_056505.2). | |
| 100 μg | |
| Protein Kinase | |
| 27148 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion