Learn More
STK32B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15438-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a serine-threonine protein kinase. Serine-threonine kinases transfer phosphate molecules to the oxygen atoms of serine and threonine. A genomic deletion affecting this gene has been associated with Ellis-van Creveld syndrome, an autosomal recessive skeletal dysplasia. Alternative splicing results in multiple transcript variants.Specifications
STK32B | |
Polyclonal | |
Unconjugated | |
STK32B | |
HSA250839, serine/threonine kinase 32B, STK32, STK32B, STKG6, YANK2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
305431, 55351, 64293 | |
-20°C | |
Liquid |
Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9JJX8, Q9NY57 | |
STK32B | |
STK32B Fusion Protein Ag4507 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Product Title
STK32B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction