missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human STK3 (MST2), GST Tag Recombinant Protein
Human STK3 (MST2), GST Tag Recombinant Protein
Brand: Invitrogen PV4805
Additional Details : Weight : 0.50000kg
Description
STK3 (MST2) is a STE kinase family member that acts upstream of the MAP kinase signaling pathways, and is known to be activated by the proapoptotic stimuli staurosporine and FAS ligand.Specifications
NP_006272.2 | |
6788 | |
10 μg | |
STE20 Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.02% Trition X-100, 0.5 mM EDTA, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: STE Group | |
Z'-LYTE™ | |
serine⁄threonine kinase 3 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
85.5 kDa | |
STK3 (MST2) | |
Human | |
GST Tag | |
Biochemical binding/kinetic, Biochemical phospho-specific activity (no antibody) | |
Dry Ice | |
STK3 | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |