Learn More
Invitrogen™ Human STK24 (MST3), GST Tag Recombinant Protein
Recombinant Protein
14370.00 DKK - 62200.00 DKK
Specifications
Accession Number | Q9Y6E0 |
---|---|
Concentration | See Label |
For Use With (Application) | Kinase Assay |
Formulation | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
Gene ID (Entrez) | 8428 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15498473
|
Invitrogen™
PR6359A |
100 ÎĽg |
14370.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10097082
|
Invitrogen™
PV4064 |
1 mg |
62200.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 ÎĽg, 10 ÎĽg and 20 ÎĽg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 ÎĽL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length
STK24 (MST3) is a serine/threonine kinase that is a member of the GCK subfamily of STE20-like proteins. The yeast 'Sterile 20' gene (STE20) functions upstream of the mitogen-activated protein kinase (MAPK) cascade. In mammals, protein kinases related to STE20 can be divided into 2 subfamilies based on their structure and regulation. Members of the PAK subfamily (see PAK3; MIM 300142) contain a C-terminal catalytic domain and an N-terminal regulatory domain that has a CDC42 (MIM 116952)-binding domain. In contrast, members of the GCK subfamily (see MAP4K2; MIM 603166), also called the Sps1 subfamily, have an N-terminal catalytic domain and a C-terminal regulatory domain without a CDC42-binding domain. STK24 belongs to the GCK subfamily of STE20-like kinases.Specifications
Q9Y6E0 | |
Kinase Assay | |
8428 | |
Human STK24 (MST3), GST Tag | |
1810013H02Rik; C76483; epididymis secretory protein Li 95; HEL-S-95; Mammalian STE20-like protein kinase 3; Mammalian STE20-like protein kinase 3 C-terminal; Mammalian STE20-like protein kinase 3 N-terminal; mammalian sterile twenty 3 kinase; Mst3; MST-3; MST3/C; MST3/N; MST3B; OTTHUMP00000018594; RGD1561742; RP11-111L24.5; serine/threonine kinase 24; serine/threonine kinase 24 (STE20 homolog, yeast); serine/threonine-protein kinase 24; Serine/threonine-protein kinase 24 12 kDa subunit; Serine/threonine-protein kinase 24 35 kDa subunit; Serine/threonine-protein kinase 24 36 kDa subunit; STE20; STE20-like kinase 3; STE20-like kinase MST3; sterile 20-like kinase 3; STK24; STK3 | |
STE20 Kinase Family | |
Human | |
GST-tag | |
-80°C, Avoid Freeze/Thaw Cycles | |
Liquid |
See Label | |
50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 | |
78.8 kDa | |
RUO - research use only | |
MST3 | |
Unconjugated | |
Recombinant | |
Full length | |
Insect cells |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.