Få mere at vide
Stiripentol-d9, TRC
Artikelnummer. 30428479
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428479 10 mg
30412694 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30428479 Leverandør Toronto Research Chemicals Leverandørnr. TRCS68682710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Stiripentol-d9
CAS 1185239-64-8
Renhedsgrad noter HPLC
Molekylær formel C14H9D9O3
Mængde 10 mg
InChI formel InChI=1S/C14H18O3/c1-14(2,3)13(15)7-5-10-4-6-11-12(8-10)17-9-16-11/h4-8,13,15H,9H2,1-3H3/i1D3,2D3,3D3
SMIL [2H]C([2H])([2H])C(C(O)C=Cc1ccc2OCOc2c1)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1-(1,3-benzodioxol-5-yl)-5,5,5-trideuterio-4,4-bis(trideuteriomethyl)pent-1-en-3-ol
Molekylvægt (g/mol) 243.35
Formel vægt 243.1821
Procent renhed >95