Få mere at vide
Stigmastanol, TRC
Artikelnummer. 30009739
Skift visning
Mængde:
50 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009739 1 g
30006917 50 mg
30011946 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009739 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6867151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 83-45-4
Molekylær formel C29 H52 O
Molekylvægt (g/mol) 416.72
Synonym Stigmastan-3-ol, (3β,5α)-, 5α-Stigmastan-3β-ol (6CI,7CI,8CI), (3β,5α)-Stigmastan-3-ol, 24α-Ethylcholestanol, 3β-Sitostanol, 5,6-Dihydro-β-sitosterol, Dihydro-β-sitosterol, Dihydrositosterin, Dihydrositosterol, Fucostanol, NSC 49081, Sitostanol, Spinastanol, Stigmastanol, β-Sitostanol, β-Sitosterol, dihydro-
IUPAC navn (3S,5S,8R,9S,10S,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMIL CC[C@H](CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)C
Tekniske data
Kemisk navn eller materiale Stigmastanol
Renhedsgrad noter HPLC
Mængde 1 g
InChI formel InChI=1S/C29H52O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h19-27,30H,7-18H2,1-6H3/t20-,21-,22+,23+,24+,25-,26+,27+,28+,29-/m1/s1
Formel vægt 416.4018
Procent renhed >95%