Få mere at vide
STF 118804
Beskrivelse
STF 118804 is a NAMPT inhibitor (nicotinamide phosphoribosyltransferase, visfatin, PBEF1); inhibits NAD+ synthesis from nicotinamide. Potently inhibits the viability of multiple B-cell acute lymphoplastic leukemia (B-ALL) cell lines (IC50 < 10 nM); induces apoptosis in MF-411 cells. Induces regression of ALL xenografts and eliminates leukemia stems cells from bone marrow in mice.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-[5-Methyl-4-[[(4-methylphenyl)sulfonyl]methyl]-2-oxazolyl]-N-(3-pyridinylmethyl)benzamide |
| CAS | 894187-61-2 |
| Analyseprocentområde | >98% |
| Mængde | 25 mg |
| Molekylær formel | C25H23N3O4S |
| Formel vægt | Observed MW: 461.53 |