Learn More
Sampling Systems™ SteriWare™ HDPE Disposable Powder Thief
BIOPHARMA
Sampling Systems™ SteriWare™ HDPE Disposable Powder Thief is ideal for taking representative samples from known locations within the bulk powder.
Brand:  Sampling Systems™ 8060H-1020
 View more versions of this product
Product Code. 11935457
  • 1340.00 DKK / Pack of 20
Only null left
Estimated Shipment: 05-08-2025
Description
  • Assembled and packed in a cleanroom
  • Individually Bagged
  • Made from FDA or EU conforming HDPE
  • BSE/TSE Free
  • Full Batch Traceability
  • Suitable for use in ATEX zones 0, 1 and 2
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • No cleaning is required before or after sampling
  • No cleaning validation (this is very expensive to set up and run)
  • Manufactured in a class 100,000 cleanroom environment (check individual sampler descriptions)
  • Peace of mind
  • Suitable for all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.)
  • Suitable for use in ATEX Zones 0, 1 and 2
  • Financial
  • Save time and money (it will cost more to clean a reusable sampler than to buy a disposable sampler)
Specifications
Specifications
HDPE
Non-sterile
1000 mm
20 mL
21 mm
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number117362 Certificate TypeCertificate of Compliance Date19/06/2024 Catalog Number8060H-1020
Lot Number110076 Certificate TypeCertificate of Compliance Date31/07/2023 Catalog Number8060H-1020
Lot Number109391 Certificate TypeCertificate of Compliance Date02/05/2023 Catalog Number8060H-1020
Lot Number102612 Certificate TypeCertificate of Compliance Date05/08/2022 Catalog Number8060H-1020
Lot Number107353 Certificate TypeDeclaration of Conformity Date05/12/2022 Catalog Number8060H-1020
Promotions
Promotions
Product Content Correction
Product Title
Sampling Systems™ SteriWare™ HDPE Disposable Powder Thief > Capacity: 20mL