Få mere at vide
Heinz Herenz Steristoppers No.14D for Glasses
Stopper nummer:
14D
Beskrivelse
Steristoppers™ are an invention of the Heinz Herenz company and have proven themselves worldwide for many years. The cellulose stoppers are ideal for sterile sealing of bacterial cultures in test tubes, Erlenmeyer flasks and culture flasks. Many important institutions all over the world rely on our in house invention, because in addition to its outstanding properties it also simplifies work in the laboratory.
- Autoclavable up to 200 °C
- Bacteria filtering
- Sterilizable
- Air permeable
- Good value for money
- Available in various sizes, shapes and packaging units. Special designs are also possible.
Please note:
- Only suitable for single use
Tekniske data
Tekniske data
| Engangs | Single-use |
| Autoklaverbar | Autoclavable |
| Farve | White |
| Diameter (metrisk) bund | 12 mm |
| Diameter (metrisk) Udvendig | 15 mm |
| Diameter (metrisk) Top | 15 mm |
| Materiale | Cellulose Fiber |
| Produkttype | Steri-stopper |
| Form | Conical |
| Sterilitet | Non-sterile |
| Vis mere |