Få mere at vide
Thermo Scientific™ Sterilin™ Water Sampling Bottles, 500mL PET Square bottle, labeled, dosed 120mg/L with Sodium Thiosulphate
GREENER_CHOICE
Artikelnummer. 13128770
Artikelnummer. 13128770
Brand: Thermo Scientific™ WSC500
Conveniently sample both chlorinated or nonchlorinated water for microbial analysis with Sterilin Water Sampling Bottles.
Conveniently sample both chlorinated or nonchlorinated water for microbial analysis with Thermo Scientific™ Sterilin Water Sampling Bottles.
  • Available manufactured using:
    • Flexible polystyrene
    • Robust non-toxic PETG
    • NEW, lightweight PRT for improved handling
  • Empty or pre-dosed with sodium thiosulphate
  • Polystyrene and PETG bottles feature color-coded labels to aid differentiation:
    • Blue: dosed
    • Green: undosed
  • Tamper evident for sample integrity
  • Lot number for complete traceability
  • Sterilized by gamma irradiation
  • Dose level 20mg/h: Suitable for neutralizing water samples with a low concentration of chlorine
  • Dose level 120mg/h: Suitable for neutralizing water samples with a high concentration of chlorine