Få mere at vide
Bel-Art™ Sterileware™ Jumbo prøveudtagningspipetter
Shop alle SP Scienceware produkter
Kapacitet (metrisk):
80 ml
180 ml
225 ml
Beskrivelse
- Reaches into container while keeping hand clear of contents
- Diameter allows for use through drum bunghole or other small openings
- Color-coded end helps prevent handling errors
- Low-viscosity liquids enter easily through the 3 mm (0.125 in.) tip hole
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) pipettes heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10^-6)
- Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Tekniske data
Tekniske data
| Produkttype | Liquid Sampler |
| Materiale | HDPE |
| Autoklaverbar | Not Autoclavable |
| Kapacitet (engelsk) | 80 mL |
| Kapacitet (metrisk) | 80 ml |
| Mængde | 50/Pk. |
| Diameter (metrisk) Udvendig | 2.1 cm |
| Længde (metrisk) | 45 cm |