Få mere at vide
Bel-Art™ Sterileware™ Tværsnits væskeprøveudtagere
Shop alle SP Scienceware produkter
Kapacitet (metrisk):
80 ml
225 ml
Beskrivelse
- Provides effective cross-sectional sampling of liquids and light creams
- Reaches bottom of container while keeping hand clear of container top
- Diameter allows for use through drum bunghole or other small openings
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) samplers heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10^-6)
- Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Tekniske data
Tekniske data
| Produkttype | Liquid Sampler |
| Materiale | HDPE |
| Autoklaverbar | Not Autoclavable |
| Kapacitet (engelsk) | 2.75 oz. |
| Kapacitet (metrisk) | 80 ml |
| Mængde | 20/Pk. |
| Diameter (metrisk) Udvendig | 2.1 cm |
| Længde (metrisk) | 45 cm |