missing translation for 'onlineSavingsMsg'
Få mere at vide

Stenbolone, TRC

Artikelnummer. 30414578
Skift visning
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30414578 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414578 Supplier Toronto Research Chemicals Supplier No. TRCS68670010MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Stenbolone
CAS 5197-58-0
Molekylær formel C20 H30 O2
Mængde 10 mg
Synonym 5alpha-Androst-1-en-3-one','17beta-hydroxy-2-methyl- (6CI,7CI,8CI), (5alpha,17beta)-17-hydroxy-2-methyl-Androst-1-en-3-one, 17beta-Hydroxy-2-methyl-5alpha-androst-1-en-3-one, NSC 34418, NSC 65890, Stenbolone, Stenebolone
InChI formel InChI=1S/C20H30O2/c1-12-11-20(3)13(10-17(12)21)4-5-14-15-6-7-18(22)19(15,2)9-8-16(14)20/h11,13-16,18,22H,4-10H2,1-3H3/t13-,14-,15-,16-,18-,19-,20-/m0/s1
SMIL CC1=C[C@@]2(C)[C@@H](CC[C@H]3[C@@H]4CC[C@H](O)[C@@]4(C)CC[C@H]23)CC1=O
IUPAC navn (5S,8R,9S,10S,13S,14S,17S)-17-hydroxy-2,10,13-trimethyl-4,5,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one
Molekylvægt (g/mol) 302.45
Formel vægt 302.2246
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.