Learn More
RETSCH Steel Parallel Section Rotor
Brand: RETSCH 22.608.0018
Additional Details : Weight : 4.30000kg
Specifications
Parallel Section Rotor | |
For grinding without heavy-metal contamination | |
For SM 300 Cutting Mill | |
Steel |