Få mere at vide
Juchheim Laborgeraete™ Steel Mortar with Locking Nut
Artikelnummer. 10483512
Diameter (metrisk) Indvendig:
20 mm
Artikelnummer. 10483512
Brand: Juchheim Laborgeraete™ 00470
Heavy duty diamond mortar for effective crushing and pulverizing in the laboratory.
  • Option of inner diameter of 20 mm, 30 mm, and 40 mm
Tekniske data
Kapacitet (metrisk) 30 mL
Slutte Hardened Steel
Diameter (metrisk) Indvendig 20 mm
Produkttype Mortar
Materiale Hardened Steel
Inkluderer Mortar with Locking Nut