Få mere at vide
Juchheim Laborgeraete™ Steel Diamond Mortar
Artikelnummer. 10559882
Diameter (metrisk) Indvendig:
20 mm
30 mm
Artikelnummer. 10559882
Brand: Juchheim Laborgeraete™ 00461
Heavy duty diamond mortar for effective crushing and pulverizing in the laboratory.
  • Option of inner diameter of 20 mm, 30 mm, and 40 mm
Tekniske data
Kapacitet (metrisk) 50 mL
Slutte Hardened Steel
Diameter (metrisk) Indvendig 30 mm
Produkttype Mortar
Materiale Hardened Steel