Få mere at vide
Stearoyl-L-carnitine-d3 Hydrochloride, TRC
Artikelnummer. 30420092
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30420092 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30420092 Leverandør Toronto Research Chemicals Leverandørnr. TRCS68651225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Stearoyl-L-carnitine-d3 Hydrochloride
CAS 2245711-27-5
Molekylær formel C25 D3 H47 N O4 . Cl
Mængde 25 mg
Synonym 1-Propanaminium','3-carboxy-N,N-dimethyl-N-(methyl-d3)-2-[(1-oxooctadecyl)oxy]-','chloride (1:1)','(2R)- (ACI), Octadecanoyl-L-carnitine-d3 HCl (N-methyl-d3), O-Octadecanoyl-L-carnitine-d3 chloride, Stearoyl-L-carnitine-d3 chloride, Stearoyl-L-carnitine-D3 (N-methyl-D3) chloride, (2R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-[(1-oxooctadecyl)oxy]-1-propanaminium chloride
InChI formel InChI=1S/C25H49NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-25(29)30-23(21-24(27)28)22-26(2,3)4;/h23H,5-22H2,1-4H3;1H/t23-;/m1./s1/i2D3;
SMIL [Cl-].[2H]C([2H])([2H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)CCCCCCCCCCCCCCCCC
IUPAC navn [(2R)-3-carboxy-2-octadecanoyloxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride
Molekylvægt (g/mol) 467.14
Formel vægt 466.3617