Få mere at vide
Stearoyl Ethanolamide, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Stearoyl Ethanolamide |
| CAS | 111-57-9 |
| Molekylær formel | C20H41NO2 |
| Mængde | 1 g |
| Synonym | N-(2-Hydroxyethyl)octadecanamide, AM 1105, Abriflo 65, Alkamide S 280, Aminon SM, Amisol SME, Ceramid, Clindrol 200MS, Comperlan HS, Cycloamide SM, Emcol 70, Loramine S 280 Lubsize K 12, Mackamide SMA, Marlamid M 18, Monamid 972, Monamid S, Monoethanolstearamide, N-(2-Hydroxyethyl)stearamide, N-Octadecanoylethanolamine, N-Stearoylethanolamine, NSC 3377, NSC 52619, Onyx Wax EL, Profan SME, Rewomid S 280, S 280, Stearamide MEA, Stearamide monoethanolamine, Stearic acid ethanolamide, Stearic acid monoethanolamide, Stearic ethanolamide, Stearic ethylolamide, Stearic monoethanolamide, Stearic monoethanolamine, Stearoylmonoethanolamide, Witcamide 70 |
| InChI formel | InChI=1S/C20H41NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(23)21-18-19-22/h22H,2-19H2,1H3,(H,21,23) |
| SMIL | CCCCCCCCCCCCCCCCCC(NCCO)=O |
| IUPAC navn | N-(2-hydroxyethyl)octadecanamide |
| Molekylvægt (g/mol) | 327.55 |
| Formel vægt | 327.3137 |