Få mere at vide
Stearidonic Acid, TRC
Artikelnummer. 30137269
Skift visning
Mængde:
0.5 mg
2.5 mg
10 mg
Pakningsstørrelse:
0.5mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30137269 10 mg -
30006341 0.5 mg 0.5mg
30007828 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137269 Leverandør Toronto Research Chemicals Leverandørnr. TRCS68645010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 20290-75-9
Molekylær formel C18 H28 O2
Molekylvægt (g/mol) 276.41
Synonym 6,9,12,15-Octadecatetraenoic acid, (6Z,9Z,12Z,15Z)-, 6,9,12,15-Octadecatetraenoic acid, (all-Z)- (8CI), (6Z,9Z,12Z,15Z)-6,9,12,15-Octadecatetraenoic acid, (6Z,9Z,12Z,15Z)-Octadecatetraenoic acid, (Z,Z,Z,Z)-6,9,12,15-Octadecatetraenoic acid, (all-Z)-6,9,12,15-Octadecatetraenoic acid, 18:4(n-3), 6,9,12,15-cis-Octadecatetraenoic acid, 6Z,9Z,12Z,15Z-Octadecatetraenoic acid, Moroctic acid, Stearidonic acid, all-cis-6,9,12,15-Octadecatetraenoic acid, δ6,9,12,15-Octadecatetraenoic acid
IUPAC navn (6Z,9Z,12Z,15Z)-octadeca-6,9,12,15-tetraenoic acid
SMIL CC\C=C/C\C=C/C\C=C/C\C=C/CCCCC(=O)O
Tekniske data
Kemisk navn eller materiale Stearidonic Acid
Mængde 10 mg
InChI formel InChI=1S/C18H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h3-4,6-7,9-10,12-13H,2,5,8,11,14-17H2,1H3,(H,19,20)/b4-3-,7-6-,10-9-,13-12-
Formel vægt 276.209