missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Stearic-D35 Acid, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Stearic-D35 Acid |
| CAS | 17660-51-4 |
| Molekylær formel | C18 D35 H O2 |
| Mængde | 10 mg |
| Synonym | Octadecanoic-D35 acid, Octadecanoic-d35 acid (9CI','ACI), Stearic-d35 acid (8CI), Stearic acid-2H35, Octadecanoic acid D35 |
| InChI formel | InChI=1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2 |
| SMIL | [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O |
| IUPAC navn | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,18,18,18-pentatriacontadeuteriooctadecanoic acid |
| Molekylvægt (g/mol) | 319.69 |
| Formel vægt | 319.4912 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?