missing translation for 'onlineSavingsMsg'
Learn More

Stearic Acid NF/FCC Powder, Macron Fine Chemicals™

Catalog Number p-3178348
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
POLYSTORMOR
STAKMOR
This item is not returnable. View return policy
57-11-4
C18H36O2
284.48
MFCD00002752
QIQXTHQIDYTFRH-UHFFFAOYSA-N
stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid
5281
CHEBI:28842
octadecanoic acid
CCCCCCCCCCCCCCCCCC(O)=O
This item is not returnable. View return policy
57-11-4
C18H36O2
284.48
MFCD00002752
QIQXTHQIDYTFRH-UHFFFAOYSA-N
stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid
5281
CHEBI:28842
octadecanoic acid
CCCCCCCCCCCCCCCCCC(O)=O
Packaging POLYSTORMOR
Quantity 500 g
Formula Weight 284.48
Grade FCC/NF
Physical Form Powder
Chemical Name or Material Stearic Acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'