missing translation for 'onlineSavingsMsg'
Learn More
Staurosporine, MedChemExpress Product Code.: 16480226

Staurosporine, MedChemExpress

Product Code. p-200098366
2 mg, 2mg
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16480226

Brand: Medchem Express HY15141

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Staurosporine is a potent, ATP-competitive and non-selective inhibitor of protein kinases with IC50s of 6 nM, 15 nM, 2 nM, and 3 nM for PKC, PKA, c-Fgr, and Phosphorylase kinase respectively. Staurosporine also inhibits TAOK2 with an IC50 of 3 ÎĽM. Staurosporine is an apoptosis inducer.

Staurosporine is a potent, ATP-competitive and non-selective inhibitor of protein kinases with IC50s of 6 nM, 15 nM, 2 nM, and 3 nM for PKC, PKA, c-Fgr, and Phosphorylase kinase respectively. Staurosporine also inhibits TAOK2 with an IC50 of 3 ÎĽM. Staurosporine is an apoptosis inducer.

Specifications

Chemical Name or Material Staurosporine
CAS 62996-74-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C28H26N4O3
Quantity 2 mg
Synonym Antibiotic AM-2282 STS AM-2282
Solubility Information DMSO : 62.5 mg/mL (133.97 mM; Need ultrasonic)
SMILES O=C(NC1)C2=C1C3=C(C4=C2C5=C(C=CC=C5)N4[C@H]6C[C@@H](NC)[C@@H](OC)[C@]7(C)O6)N7C8=CC=CC=C83
Molecular Weight (g/mol) 466.53
Formula Weight 466.53
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Staurosporine, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.