missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Staufen Recombinant Rabbit Monoclonal Antibody (K01_3L68)

Artikelnummer. 30826673
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30826673 50 μL
30827064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30826673 Leverandør Invitrogen™ Leverandørnr. MA565861

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: DQQSTEMPRT GNGP.

STAU1 is a member of a family of double-stranded RNA (dsRNA)-binding proteins involved in the transport and/or localization of mRNAs to different subcellular compartments and/or organelles. These proteins are characterized by the presence of multiple dsRNA-binding domains which are required to bind RNAs having double-stranded secondary structures. The human STAU1 also contains a microtubule- binding domain similar to that of microtubule-associated protein 1B, and binds tubulin. STAU1 has been shown to be present in the cytoplasm in association with the rough endoplasmic reticulum (RER), implicating this protein in the transport of mRNA via the microtubule network to the RER. STAU1 is also known to interact with influenza ribonucleoproteins NS1, NP, and PA and is required for efficient viral replication.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Staufen
Applikationer Immunohistochemistry (Paraffin), Western Blot
Klassifikation Recombinant Monoclonal
Klon K01_3L68
Koncentration 1 mg/mL
Konjugeret Unconjugated
Formulering 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene Stau1
Genadgangsnr. O95793, Q9Z108
Gene Alias 5830401L18Rik; AW549911; C85792; Double-stranded RNA-binding protein Staufen homolog 1; PPP1R150; protein phosphatase 1, regulatory subunit 150; RP3-470L14.2; STAU; Stau1; staufen (RNA binding protein) homolog 1 (Drosophila); staufen double-stranded RNA binding protein 1; staufen RNA binding protein homolog 1; staufen, RNA binding protein, homolog 1
Gensymboler Stau1
Værtsarter Rabbit
Immunogen A synthetic peptide of human Staufen (aa 557-570).
Oprensningsmetode Protein A/G
Mængde 50 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 20853, 6780, 84496
Målarter Human, Mouse, Rat
Indhold og opbevaring Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.