missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Starch

Soluble GR for analysis ISO

Brand:  Merck Millipore 1012521000

Additional Details : CAS Number : 9005-84-9 Weight : 1.00000kg

Quantity 1 kg

 View more versions of this product

Product Code. 10215030

  • 2145.00 DKK / 1kg


Estimated Shipment
10-05-2021
Add to basket

Description

Description

  • Carbohydrate
  • Starch is generally composed of two molecule types: amylose and amylopectin. Starch and modified starches are important excipients. They act as fillers/binders.
  • Appearance (color): white
  • Appearance (description): fine powder
  • pH-value (2 %; water): 6.0 - 7.5
  • Sensitivity: passes test
  • Reducing matter (as maltose): max 0.7
  • Sulfated ash: max 0.4
  • Loss on drying (105 °C, 2 h): max 10
  • Suitability as enzyme substrate (for amylases): passes test
    Iso reagent
Specifications

Chemical Identifiers

1 S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20 H,1-2H2/t3-,4-,5-,6+,7-,8-,9-,10-,11+,12-/m1/s1
6.0-7.5 (25°C, 20 g/L in H2O)
Amylum, Potato starch for determination of diastase
GUBGYTABKSRVRQ-ASMJPISFSA-N
50 g/L

Specifications

Amylum, Potato starch for determination of diastase
GUBGYTABKSRVRQ-ASMJPISFSA-N
6.0-7.5 (25°C, 20 g/L in H2O)
1 S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20 H,1-2H2/t3-,4-,5-,6+,7-,8-,9-,10-,11+,12-/m1/s1
50 g/L
1 kg
Videos
SDS
Documents