Få mere at vide
Stanozolol-d3, TRC
Artikelnummer. 30425993
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30425993 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30425993 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6845022.5MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Stanozolol-d3
CAS 792214-66-5
Molekylær formel C21 D3 H29 N2 O
Mængde 2.5 mg
Synonym 1'H-Androstano[3,2-c]pyrazol-17-ol','17-(methyl-d3)-','(5α,17β)- (9CI), Stanozolol D3 (17-methyl D3), Stanozolol-d3, (5α,17β)-17-(Methyl-d3)-2'H-androst-2-eno[3,2-c]pyrazol-17-ol, 17-(Methyl-d3)-5α-androstano[3,2-c]pyrazol-17β-ol, 17β-Hydroxy-17-(methyl-d3)-5α-androstano[3,2-c]pyrazole
InChI formel InChI=1S/C21H32N2O/c1-19-11-13-12-22-23-18(13)10-14(19)4-5-15-16(19)6-8-20(2)17(15)7-9-21(20,3)24/h12,14-17,24H,4-11H2,1-3H3,(H,22,23)/t14-,15+,16-,17-,19-,20-,21-/m0/s1/i3D3
SMIL [2H]C([2H])([2H])[C@]1(O)CC[C@H]2[C@@H]3CC[C@H]4Cc5[nH]ncc5C[C@]4(C)[C@H]3CC[C@]12C
Molekylvægt (g/mol) 331.51
Formel vægt 331.27