Learn More
Macherey-Nagel™ Silica 60 Standard Adsorbent for Column Chromatography
Use silca 60 for standard separations. Silica 60 Standard Adsorbent for Column Chromatography comes in a variety of particle sizes and package sizes.
Brand:  Macherey-Nagel™ 815360.1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10566881
  • 387.00 DKK / 1kg
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Description
Description
  • Pore size ∼ 60 Å
  • Pore volume ∼ 0.75 mL/g
  • Spec. surface BET ∼ 500 m2/g;
  • Highly porous
  • Amorphous silicic acid in the form of hard, opalescent particles
  • Prepared by precipitation of water glass with sulfuric acid
Specifications
Specifications
0.5 to 1.0 mm
Column Chromatography
1 kg
∼500 m2/g
Column Chromatography
∼60 Å
Plastic Container
Silica
Standard Silica 60 Adsorbent